Learn More
3-(2-Thienyl)-L-alanine, 95%
CAS: 22951-96-8 | C7H9NO2S | 171.214 g/mol
Supplier: Thermo Scientific Chemicals H6331603
Description
3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
| 255°C to 263°C (decomposition) | |
| 22951-96-8 | |
| C7H9NO2S | |
| 82872 | |
| Slightly soluble in water. | |
| C1=CSC(=C1)CC(C(=O)O)N | |
| 1 g | |
| 146719 | |
| 95% |
| 3-(2-Thienyl)-L-alanine | |
| White | |
| MFCD00064335 | |
| 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715 | |
| WTOFYLAWDLQMBZ-LURJTMIESA-N | |
| (2S)-2-amino-3-thiophen-2-ylpropanoic acid | |
| 171.214 | |
| 171.21 | |
| Solid |
Chemical Identifiers
| 22951-96-8 | |
| 171.214 | |
| WTOFYLAWDLQMBZ-LURJTMIESA-N | |
| 146719 | |
| C1=CSC(=C1)CC(C(=O)O)N |
| C7H9NO2S | |
| MFCD00064335 | |
| 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715 | |
| (2S)-2-amino-3-thiophen-2-ylpropanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only