Learn More
3-(2-Thienyl)-L-alanine, 95%
CAS: 22951-96-8 | C7H9NO2S | 171.214 g/mol
$298.34 - $1165.10
Chemical Identifiers
| CAS | 22951-96-8 |
|---|---|
| Molecular Formula | C7H9NO2S |
| Molecular Weight (g/mol) | 171.214 |
| MDL Number | MFCD00064335 |
| InChI Key | WTOFYLAWDLQMBZ-LURJTMIESA-N |
| Synonym | 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715 |
| PubChem CID | 146719 |
| IUPAC Name | (2S)-2-amino-3-thiophen-2-ylpropanoic acid |
| SMILES | C1=CSC(=C1)CC(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6331603
|
Thermo Scientific Chemicals
H6331603 |
1 g |
Each for $298.34
|
|
|||||
|
AAH6331606
|
Thermo Scientific Chemicals
H6331606 |
5 g |
Each for $1,165.10
|
|
|||||
Description
3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 22951-96-8 | |
| 171.214 | |
| WTOFYLAWDLQMBZ-LURJTMIESA-N | |
| 146719 | |
| C1=CSC(=C1)CC(C(=O)O)N |
| C7H9NO2S | |
| MFCD00064335 | |
| 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715 | |
| (2S)-2-amino-3-thiophen-2-ylpropanoic acid |
Specifications
| 255°C to 263°C (decomposition) | |
| White | |
| MFCD00064335 | |
| 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715 | |
| WTOFYLAWDLQMBZ-LURJTMIESA-N | |
| (2S)-2-amino-3-thiophen-2-ylpropanoic acid | |
| 171.214 | |
| 171.21 | |
| Solid |
| 22951-96-8 | |
| C7H9NO2S | |
| 82872 | |
| Slightly soluble in water. | |
| C1=CSC(=C1)CC(C(=O)O)N | |
| 1 g | |
| 146719 | |
| 95% | |
| 3-(2-Thienyl)-L-alanine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only