missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyridyl)propionic acid, 97%

Catalog No. p-7039386
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5861903 1 g
AAH5861906 5 g
2 options

Catalog No. AAH5861903

Supplier: Thermo Scientific Chemicals H5861903

Only null left

CAS: 15197-75-8 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15197-75-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00995159
InChI Key AZYKGQOIAGPVCK-UHFFFAOYSA-N
Synonym 3-pyridin-2-yl propanoic acid, 3-2-pyridyl propionic acid, 3-pyridin-2-yl-propionic acid, 2-pyridinepropanoic acid, 3-2-pyridinyl propanoic acid, pyridine-2-propionic acid, 2-3-pyridyl propionic acid, 3-2-pyridyl propanoic acid, 3-2-pyridyl-propionic acid, 2-pyridinepropanoicacid
PubChem CID 564292
IUPAC Name 3-pyridin-2-ylpropanoic acid
SMILES C1=CC=NC(=C1)CCC(=O)O

Specifications

Quantity 1 g
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 3-(2-Pyridyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only