CAS RN 15197-75-8
IUPAC Name:
3-(pyridin-2-yl)propanoic acid
Synonyms:
3-(pyridin-2-yl)propanoic acid
Molecular Weight (g/mol):
151.17
Molecular Formula:
C8H9NO2
InChi Key:
AZYKGQOIAGPVCK-UHFFFAOYSA-N
SMILES:
OC(=O)CCC1=CC=CC=N1
1
–
1
of
1
results
1
Promotions Available
3-(2-Pyridyl)propionic acid, 97%
3-(2-Pyridyl)propionic acid, 97%
CAS: 15197-75-8 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.165 MDL Number: MFCD00995159 InChI Key: AZYKGQOIAGPVCK-UHFFFAOYSA-N Synonym: 3-pyridin-2-yl propanoic acid,3-2-pyridyl propionic acid,3-pyridin-2-yl-propionic acid,2-pyridinepropanoic acid,3-2-pyridinyl propanoic acid,pyridine-2-propionic acid,2-3-pyridyl propionic acid,3-2-pyridyl propanoic acid,3-2-pyridyl-propionic acid,2-pyridinepropanoicacid PubChem CID: 564292 IUPAC Name: 3-pyridin-2-ylpropanoic acid SMILES: C1=CC=NC(=C1)CCC(=O)O