missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrate, 97+%

Catalog No. p-3737961
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC171010010 1 g Glass bottle
AC171010050 5 g Glass bottle
AC171010250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC171010010 Supplier Thermo Scientific Chemicals Supplier No. 171010010
Only null left

CAS: 1264198-47-1 | C20H12N4Na2O6S2 | 514.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1264198-47-1 , 28048-33-1
Molecular Formula C20H12N4Na2O6S2
Molecular Weight (g/mol) 514.44
MDL Number MFCD00717621
InChI Key DDIHLFCSEHAOJZ-UHFFFAOYSA-L
Synonym 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate, disodium ferrozine 2-hydrate, 3-2-pyridyl-5,6-bis 4-sulfophenyl-1,2,4-triazine disodium salt hydrate
PubChem CID 91871971
IUPAC Name disodium;4-[3-pyridin-2-yl-6-(4-sulfonatophenyl)-1,2,4-triazin-5-yl]benzenesulfonate;hydrate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=C(N=C(N=N1)C1=CC=CC=N1)C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300.0°C
Color Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 1 g
Solubility Information (5% acetate-buffer pH 5.5) Clear yellow
Formula Weight 514.45
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only