accessibility menu, dialog, popup

UserName

NNNNSOOOSOOONaNa

CAS RN 1264198-47-1

IUPAC Name: disodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate
Synonyms: ferrozine
Molecular Weight (g/mol): 514.44
Molecular Formula: C20H12N4Na2O6S2
InChi Key: DDIHLFCSEHAOJZ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=C(N=C(N=N1)C1=CC=CC=N1)C1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)