missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole, 99%, Thermo Scientific Chemicals

Catalog No. p-7033535
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5066503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5066503 Supplier Thermo Scientific Chemicals Supplier No. H5066503

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36770-50-0
Molecular Formula C12H9N5
Molecular Weight (g/mol) 223.239
MDL Number MFCD00107797
InChI Key WOFXUCJTFGKKIF-UHFFFAOYSA-N
Synonym 3-2-pyridyl-5-4-pyridyl-1,2,4-triazole, 2-3-pyridin-4-yl-1h-1,2,4-triazol-5-yl pyridine, 3-2-pyridyl-5-4-pyridyl-1h-1,2,4-triazole, 2-3-4-pyridyl-1h-1,2,4-triazol-5-yl pyridine, 2-5-pyridin-4-yl-1h-1,2,4-triazol-3-yl pyridine, 2-5-4-pyridyl-1h-1,2,4-triazol-3-yl pyridine, acmc-1ah5x, 3-2'-pyridyl-5-4-pyridyl-1,2,4-triazole
PubChem CID 2803172
IUPAC Name 2-(3-pyridin-4-yl-1H-1,2,4-triazol-5-yl)pyridine
SMILES C1=CC=NC(=C1)C2=NC(=NN2)C3=CC=NC=C3

Specifications

Quantity 1 g
Formula Weight 223.24
Percent Purity 99%
Chemical Name or Material 3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only