CAS RN 36770-50-0
IUPAC Name:
2-[3-(pyridin-4-yl)-1H-1,2,4-triazol-5-yl]pyridine
Synonyms:
2-[5-(pyridin-4-yl)-2H-1,2,4-triazol-3-yl]pyridine
Molecular Weight (g/mol):
223.24
Molecular Formula:
C12H9N5
InChi Key:
WOFXUCJTFGKKIF-UHFFFAOYSA-N
SMILES:
N1N=C(N=C1C1=CC=CC=N1)C1=CC=NC=C1
1
–
1
of
1
results
1
Promotions Available
3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole, 99%, Thermo Scientific Chemicals
3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole, 99%, Thermo Scientific Chemicals
CAS: 36770-50-0 Molecular Formula: C12H9N5 Molecular Weight (g/mol): 223.239 MDL Number: MFCD00107797 InChI Key: WOFXUCJTFGKKIF-UHFFFAOYSA-N Synonym: 3-2-pyridyl-5-4-pyridyl-1,2,4-triazole,2-3-pyridin-4-yl-1h-1,2,4-triazol-5-yl pyridine,3-2-pyridyl-5-4-pyridyl-1h-1,2,4-triazole,2-3-4-pyridyl-1h-1,2,4-triazol-5-yl pyridine,2-5-pyridin-4-yl-1h-1,2,4-triazol-3-yl pyridine,2-5-4-pyridyl-1h-1,2,4-triazol-3-yl pyridine,acmc-1ah5x,3-2'-pyridyl-5-4-pyridyl-1,2,4-triazole PubChem CID: 2803172 IUPAC Name: 2-(3-pyridin-4-yl-1H-1,2,4-triazol-5-yl)pyridine SMILES: C1=CC=NC(=C1)C2=NC(=NN2)C3=CC=NC=C3