missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$143.54 - $143.54
Specifications
| CAS | 870774-29-1 |
|---|---|
| Color | White-Yellow |
| Molecular Formula | C16H13BO2 |
| MDL Number | MFCD16293738 |
| InChI Key | WLKQDOGZCIYEOM-UHFFFAOYSA-N |
Specifications
| 870774-29-1 | |
| C16H13BO2 | |
| WLKQDOGZCIYEOM-UHFFFAOYSA-N | |
| 248.09 |
| White-Yellow | |
| MFCD16293738 | |
| 45382255 | |
| Crystalline Powder |