missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. N10091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10091G Supplier TCI America Supplier No. N10091G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-(2-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 870774-29-1
Color White-Yellow
Molecular Formula C16H13BO2
Quantity 1 g
MDL Number MFCD16293738
Synonym 3-2-naphthyl phenylboronic acid, 3-naphthalen-2-yl phenyl boronic acid, 3-2-naphthyl benzeneboronic acid, 3-naphthalene-2-yl phenylboronic acid, boronic acid, 3-2-naphthalenyl phenyl, 3-naphthalen-2-yl phenylboronic acid, 3-2-naphthyl phenylboronicacid, 3-2-naphthyl phenyl boronic acid
InChI Key WLKQDOGZCIYEOM-UHFFFAOYSA-N
SMILES OB(O)C1=CC(=CC=C1)C1=CC=C2C=CC=CC2=C1
IUPAC Name [3-(naphthalen-2-yl)phenyl]boronic acid
Molecular Weight (g/mol) 248.09
PubChem CID 45382255
Formula Weight 248.09
Physical Form Crystalline Powder
Show More Show Less