missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-L-alanine 97.0+%, TCI America™

Catalog No. N06461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06461G 1 g
N06465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06461G Supplier TCI America Supplier No. N06461G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58438-03-2
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD00066087
InChI Key JPZXHKDZASGCLU-LBPRGKRZSA-N
Synonym 3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid
PubChem CID 185915
IUPAC Name (2S)-2-amino-3-(naphthalen-2-yl)propanoic acid
SMILES N[C@@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 1 g
Formula Weight 215.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Naphthyl)-L-alanine