missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Naphthyl)-L-alanine Hydrochloride 98.0+%, TCI America™
$253.51 - $764.27
Chemical Identifiers
| CAS | 122745-12-4 |
|---|---|
| Molecular Formula | C13H14ClNO2 |
| Molecular Weight (g/mol) | 251.71 |
| MDL Number | MFCD03095595 |
| InChI Key | QXEYSNIVQNSZGX-YDALLXLXSA-N |
| Synonym | (S)-alpha-Amino-2-naphthalenepropionic Acid Hydrochloride |
| PubChem CID | 44630411 |
| IUPAC Name | (2S)-2-amino-3-naphthalen-2-ylpropanoic acid;hydrochloride |
| SMILES | C1=CC=C2C=C(C=CC2=C1)CC(C(=O)O)N.Cl |
Chemical Identifiers
| 122745-12-4 | |
| 251.71 | |
| QXEYSNIVQNSZGX-YDALLXLXSA-N | |
| 44630411 | |
| C1=CC=C2C=C(C=CC2=C1)CC(C(=O)O)N.Cl |
| C13H14ClNO2 | |
| MFCD03095595 | |
| (S)-alpha-Amino-2-naphthalenepropionic Acid Hydrochloride | |
| (2S)-2-amino-3-naphthalen-2-ylpropanoic acid;hydrochloride |
Specifications
| 122745-12-4 | |
| MFCD03095595 | |
| (S)-alpha-Amino-2-naphthalenepropionic Acid Hydrochloride | |
| C1=CC=C2C=C(C=CC2=C1)CC(C(=O)O)N.Cl | |
| 251.71 | |
| 251.71 | |
| Crystalline Powder |
| C13H14ClNO2 | |
| 1 g | |
| QXEYSNIVQNSZGX-YDALLXLXSA-N | |
| (2S)-2-amino-3-naphthalen-2-ylpropanoic acid;hydrochloride | |
| 44630411 | |
| ≥98.0% (HPLC,T) | |
| 3-(2-Naphthyl)-L-alanine Hydrochloride |
Safety and Handling
TSCA : No