CAS RN 122745-12-4
CAS RN 122745-12-4
IUPAC Name:
2-amino-3-(naphthalen-2-yl)propanoic acid hydrochloride
Synonyms:
β-(2-naphthyl)-alanine hydrochloride
Molecular Weight (g/mol):
251.71
Molecular Formula:
C13H14ClNO2
InChi Key:
QXEYSNIVQNSZGX-UHFFFAOYNA-N
SMILES:
Cl.NC(CC1=CC2=CC=CC=C2C=C1)C(O)=O
1
–
1
of
1
results
1
3-(2-Naphthyl)-L-alanine Hydrochloride 98.0+%, TCI Americaâ„¢
CAS: 122745-12-4 Molecular Formula: C13H14ClNO2 Molecular Weight (g/mol): 251.71 MDL Number: MFCD03095595 InChI Key: QXEYSNIVQNSZGX-YDALLXLXSA-N Synonym: (S)-alpha-Amino-2-naphthalenepropionic Acid Hydrochloride PubChem CID: 44630411 IUPAC Name: (2S)-2-amino-3-naphthalen-2-ylpropanoic acid;hydrochloride SMILES: C1=CC=C2C=C(C=CC2=C1)CC(C(=O)O)N.Cl