missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-L-alanine, 97%

Catalog No. AAH6335403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6335403 1 g
AAH6335406 5 g
AAH6335414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6335403 Supplier Thermo Scientific Chemicals Supplier No. H6335403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 58438-03-2 | C13H13NO2 | 215.25 g/mol

3-(2-Naphthyl)-L-alanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Naphthyl)-L-alanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1mol/L NaOH.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 58438-03-2
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD00066087
InChI Key JPZXHKDZASGCLU-LBPRGKRZSA-N
Synonym 3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid
PubChem CID 185915
IUPAC Name (2S)-2-amino-3-naphthalen-2-ylpropanoic acid
SMILES N[C@@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O

Specifications

Beilstein 2726272
Solubility Information Soluble in 1mol/L NaOH.
Quantity 1 g
Formula Weight 215.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(2-Naphthyl)-L-alanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only