missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. M26735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26735G 5 g
M267325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26735G Supplier TCI America Supplier No. M26735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6342-77-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00002772
InChI Key XSZSNLOPIWWFHS-UHFFFAOYSA-N
Synonym 3-2-methoxyphenyl propanoic acid, 3-2-methoxyphenyl propionic acid, 2-methoxyhydrocinnamic acid, benzenepropanoic acid, 2-methoxy, o-methoxyhydrocinnamic acid, 3-o-methoxyphenyl propionic acid, 2-methoxy-benzenepropanoic acid, beta-o-methoxyphenyl propionic acid, acmc-209nfi, maybridge1_000611
PubChem CID 80652
IUPAC Name 3-(2-methoxyphenyl)propanoic acid
SMILES COC1=CC=CC=C1CCC(=O)O

Specifications

Melting Point 88°C
Color White-Yellow
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Methoxyphenyl)propionic Acid