missing translation for 'onlineSavingsMsg'
Learn More

Kaolin, Powder, FCC, Spectrum™ Chemical

Catalog No. p-7157643
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603616 500 g Poly Pail
18603618 2.5 kg Poly Pail
18603617 6 x 500 g Poly Pail
18603619 4 x 2.5 kg Poly Pail
18603620 12 kg Poly Pail
18603621 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18603616 Supplier Spectrum Chemical Mfg Cor Supplier No. K1004500GM

K1004, 1332-58-7

Spectrum™ Chemical Kaolin, Powder, FCC is used as a whitener in toothpaste. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
MDL Number MFCD00062311
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
IUPAC Name dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 500 g
Grade FCC
Loss on Ignition 15%
Packaging Poly Pail
Chemical Name or Material Kaolin, Powder