missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)propanoic acid, 98%

Catalog No. p-2698989
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330100010 1 g Glass bottle
AC330100100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC330100010 Supplier Thermo Scientific Chemicals Supplier No. 330100010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 935-13-7 | C7H8O3 | 140.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 935-13-7
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
InChI Key XLTJXJJMUFDQEZ-UHFFFAOYSA-N
Synonym 3-furan-2-yl propanoic acid, 3-2-furyl propanoic acid, 2-furanpropanoic acid, 2-furanpropionic acid, 3-2-furyl propionic acid, 3-furan-2-yl-propionic acid, furan-2-propionic acid, 3-2-furyl propoinic acid, furfuryl-essigsaure, furfurylacetic acid
PubChem CID 70286
IUPAC Name 3-(furan-2-yl)propanoic acid
SMILES C1=COC(=C1)CCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 56°C to 59°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol,acetone,ethylether,chlorofor
Formula Weight 140.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Furyl)propanoic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 213-298-4

RUO – Research Use Only