missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC46101DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC46101DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

35461-99-5
188.182
RQVVFGRDMHDHNI-UHFFFAOYSA-N
5037933
C1=CC(=CC(=C1)C(=O)O)C2=CC=CO2
C11H8O3
MFCD02690979
3-furan-2-yl benzoic acid, 3-2-furyl benzoic acid, 3-fur-2-ylbenzoic acid, 3-furan-2-yl-benzoic acid, m-2-furyl benzoic acid, benzoic acid, 3-2-furanyl
3-(furan-2-yl)benzoic acid

Specifications

3-(2-Furyl)benzoic acid
96.5
97%
C11H8O3
MFCD02690979
RQVVFGRDMHDHNI-UHFFFAOYSA-N
3-(furan-2-yl)benzoic acid
5037933
97%
35461-99-5
100.0
Amber Glass Bottle
1g
3-furan-2-yl benzoic acid, 3-2-furyl benzoic acid, 3-fur-2-ylbenzoic acid, 3-furan-2-yl-benzoic acid, m-2-furyl benzoic acid, benzoic acid, 3-2-furanyl
C1=CC(=CC(=C1)C(=O)O)C2=CC=CO2
188.182
188.18
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions