CAS RN 35461-99-5
CAS RN 35461-99-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(2-Furyl)benzoic acid, 97%, Thermo Scientific™
CAS: 35461-99-5 Molecular Formula: C11H8O3 Molecular Weight (g/mol): 188.182 MDL Number: MFCD02690979 InChI Key: RQVVFGRDMHDHNI-UHFFFAOYSA-N Synonym: 3-furan-2-yl benzoic acid,3-2-furyl benzoic acid,3-fur-2-ylbenzoic acid,3-furan-2-yl-benzoic acid,m-2-furyl benzoic acid,benzoic acid, 3-2-furanyl PubChem CID: 5037933 IUPAC Name: 3-(furan-2-yl)benzoic acid SMILES: C1=CC(=CC(=C1)C(=O)O)C2=CC=CO2