missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Dimethylamino)ethylcarbamoyl]benzeneboronic acid, 96%, Thermo Scientific Chemicals

Catalog No. p-7034799
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5249803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5249803 Supplier Thermo Scientific Chemicals Supplier No. H5249803

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 850567-31-6
Molecular Formula C11H17BN2O3
Molecular Weight (g/mol) 236.078
MDL Number MFCD04115706
InChI Key WGZQGJKHTBTXLD-UHFFFAOYSA-N
Synonym 3-2-dimethylamino ethylcarbamoyl phenylboronic acid, 3-2-dimethylamino ethyl carbamoyl phenyl boronic acid, 3-2-dimethylaminoethylaminocarbonyl benzeneboronic acid, 3-2-dimethylamino ethyl carbamoyl phenylboronic acid, acmc-209q0i, 3-2-dimethylamino ethyl carbamoyl-phenyl boronic acid, 3-2-dimethylamino ethyl carbamoyl phenyl boronicacid, 3-2-n,n-dimethylaminoethylaminocarbonyl benzene boronic acid
PubChem CID 44119531
IUPAC Name [3-[2-(dimethylamino)ethylcarbamoyl]phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)NCCN(C)C)(O)O

Specifications

Melting Point 42°C to 45°C
Quantity 1 g
Formula Weight 236.08
Percent Purity 96%
Chemical Name or Material 3-[2-(Dimethylamino)ethylcarbamoyl]benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only