accessibility menu, dialog, popup

UserName

BONHNCH3CH3HOOH

CAS RN 850567-31-6

IUPAC Name: (3-{[2-(dimethylamino)ethyl]carbamoyl}phenyl)boronic acid
Synonyms: 3-{[2-(dimethylamino)ethyl]carbamoyl}phenylboronic acid
Molecular Weight (g/mol): 236.08
Molecular Formula: C11H17BN2O3
InChi Key: WGZQGJKHTBTXLD-UHFFFAOYSA-N
SMILES: CN(C)CCNC(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results