missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chlorophenyl)propionic acid, 98+%

Catalog No. p-7046620
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1530603 1 g
AAL1530606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1530603 Supplier Thermo Scientific Chemicals Supplier No. L1530603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1643-28-3 | C9H9ClO2 | 184.619 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1643-28-3
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00016547
InChI Key KZMDFTFGWIVSNQ-UHFFFAOYSA-N
Synonym 3-2-chlorophenyl propionic acid, 3-2-chlorophenyl propanoic acid, benzenepropanoic acid, 2-chloro, 2-chlorobenzenepropanoic acid, 2-chloro-benzenepropanoic acid, zlchem 694, pubchem19768, acmc-209dqy, 2-chlorohydrocinnamic acid, ksc494i2t
PubChem CID 95676
IUPAC Name 3-(2-chlorophenyl)propanoic acid
SMILES C1=CC=C(C(=C1)CCC(=O)O)Cl

Specifications

Melting Point 94°C to 97°C
Boiling Point 171°C (10 mmHg)
Quantity 1 g
Beilstein 1866789
Formula Weight 184.62
Percent Purity ≥98%
Chemical Name or Material 3-(2-Chlorophenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only