accessibility menu, dialog, popup

UserName

CAS RN 1643-28-3

IUPAC Name: 3-(2-chlorophenyl)propanoic acid
Synonyms: 3-(2-chlorophenyl)propanoic acid
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: KZMDFTFGWIVSNQ-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=CC=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results