missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloroethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one 98.0+%, TCI America™

Catalog No. p-7125614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27165G 5 g
C271625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27165G Supplier TCI America Supplier No. C27165G
Only null left

Chemical Identifiers

CAS 41078-70-0
Molecular Formula C11H11ClN2O
Molecular Weight (g/mol) 222.672
MDL Number MFCD03085747
InChI Key LFTGLYCNMGGMKL-UHFFFAOYSA-N
PubChem CID 817866
IUPAC Name 3-(2-chloroethyl)-2-methylpyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(C(=O)N2C=CC=CC2=N1)CCCl

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 5 g
Formula Weight 222.67
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Chloroethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one