missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 99%

Catalog No. AAL1009218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1009218 50 g
AAL1009209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1009218 Supplier Thermo Scientific Chemicals Supplier No. L1009218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3919-74-2 | C11H7ClFNO3 | 255.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3919-74-2
Molecular Formula C11H7ClFNO3
Molecular Weight (g/mol) 255.63
MDL Number MFCD00055659
InChI Key REIPZLFZMOQHJT-UHFFFAOYSA-N
Synonym 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, fcimic acid, 4-isoxazolecarboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 3-2-dihloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxic acid, pubchem8772, acmc-1ckjh, 3-2-chloro-6-fluorophenyl 5-methylisoxazole-4-carboxylic acid
PubChem CID 77520
IUPAC Name 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=C(F)C=CC=C1Cl

Specifications

Melting Point 201°C to 203°C
Quantity 50 g
Beilstein 806523
Formula Weight 255.63
Percent Purity 99%
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-486-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only