missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Bromophenyl)-1H-pyrazole, 97%

Catalog No. AAH3440609
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3440609 10 g
AAH3440603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3440609 Supplier Thermo Scientific Chemicals Supplier No. H3440609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114382-20-6 | C9H7BrN2 | 223.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114382-20-6
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.073
MDL Number MFCD01940435
InChI Key VDHTYVJAQNZWTD-UHFFFAOYSA-N
Synonym 3-2-bromophenyl-1h-pyrazole, 3-2-bromophenyl pyrazole, 5-2-bromophenyl-1h-pyrazole, 1h-pyrazole,3-2-bromophenyl, 3-2-bromo-phenyl-1h-pyrazole, 3-2-bromophenyl-2h-pyrazole, 3-o-bromophenylpyrazole, acmc-1c4z7
PubChem CID 2735612
IUPAC Name 5-(2-bromophenyl)-1H-pyrazole
SMILES C1=CC=C(C(=C1)C2=CC=NN2)Br

Specifications

Melting Point 130°C to 132°C
Quantity 10 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 3-(2-Bromophenyl)-1H-pyrazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only