missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Benzothiazolylthio)propionic Acid 98.0+%, TCI America™

Catalog No. p-7119370
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B194225G 25 g
B1942500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B194225G Supplier TCI America Supplier No. B194225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4767-00-4
Molecular Formula C10H9NO2S2
Molecular Weight (g/mol) 239.31
MDL Number MFCD00022878
InChI Key DXSBAOMLHPFLMW-UHFFFAOYSA-N
PubChem CID 224561
IUPAC Name 3-(1,3-benzothiazol-2-ylsulfanyl)propanoic acid
SMILES OC(=O)CCSC1=NC2=CC=CC=C2S1

Specifications

Melting Point 149°C
Quantity 25 g
Formula Weight 239.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Benzothiazolylthio)propionic Acid
EINECSNumber (5)-5989
TSCA No