missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propyltrimethoxysilane 97.0+%, TCI America™

Catalog No. p-7117051
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A077425G 25 g
A0774100G 100 g
A0774500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A077425G Supplier TCI America Supplier No. A077425G
Only null left

Chemical Identifiers

CAS 1760-24-3
Molecular Formula C8H22N2O3Si
Molecular Weight (g/mol) 222.36
MDL Number MFCD00008173
InChI Key PHQOGHDTIVQXHL-UHFFFAOYSA-N
Synonym n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane
PubChem CID 15659
IUPAC Name N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine
SMILES CO[Si](CCCNCCN)(OC)OC

Specifications

Color Yellow
Boiling Point 146°C
Quantity 25 g
Formula Weight 222.36
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(2-Aminoethylamino)propyltrimethoxysilane
EINECSNumber (2)-2083
RTECSNumber KV7400000
TSCA Yes