missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Aminoethylamino)propyltrimethoxysilane, 96%
Description
3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.
Solubility
Miscible with toluene.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and water.
3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.
Solubility
Miscible with toluene.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and water.
Specifications
Specifications
| Boiling Point | 261°C to 263°C |
| Quantity | 500 g |
| UN Number | UN2735 |
| Beilstein | 636230 |
| Flash Point | 136°C (276°F) |
| Solubility Information | Miscible with toluene. |
| Formula Weight | 222.36 |
| Percent Purity | 96% |
| Odor | Ammonia-like |
| Refractive Index | 1.4435 |
| Show More |
Safety and Handling
| Hazard Category | H315-H317-H318 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
| EINECSNumber | 217-164-6 |
| RTECSNumber | KV7400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only