missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)pyridine 98.0+%, TCI America™

Catalog No. A13365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13365G 5 g
A133625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13365G Supplier TCI America Supplier No. A13365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20173-24-4
Molecular Formula C7H11N2
Molecular Weight (g/mol) 123.18
MDL Number MFCD00191604
InChI Key NAHHNSMHYCLMON-UHFFFAOYSA-O
Synonym 3-2-aminoethyl pyridine, 2-pyridin-3-yl ethanamine, 3-aminoethylpyridine, 3-pyridineethanamine, 2-pyridin-3-yl ethan-1-amine, 2-3-pyridyl ethylamine, 3-pyridineethaneamine, 3-pyridine ethanamine, 2-pyridin-3-yl-ethylamine, 2-3-pyridinyl ethanamine
PubChem CID 854051
IUPAC Name 2-(pyridin-3-yl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=CN=C1

Specifications

Color Yellow
Boiling Point 128°C
Quantity 5 g
UN Number 2735
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(2-Aminoethyl)pyridine