accessibility menu, dialog, popup

UserName

CAS RN 20173-24-4

IUPAC Name: 2-(pyridin-3-yl)ethan-1-aminium
Synonyms: 2-(pyridin-3-yl)ethanaminium
Molecular Weight (g/mol): 123.18
Molecular Formula: C7H11N2
InChi Key: NAHHNSMHYCLMON-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=CN=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)