missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)pyridine, 98%

Catalog No. aal1631103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1631103 1 g
AAL1631106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1631103 Supplier Thermo Scientific Chemicals Supplier No. L1631103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20173-24-4 | C7H11N2 | 123.18 g/mol

3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.

Chemical Identifiers

CAS 20173-24-4
Molecular Formula C7H11N2
Molecular Weight (g/mol) 123.18
MDL Number MFCD00191604
InChI Key NAHHNSMHYCLMON-UHFFFAOYSA-O
Synonym 3-2-aminoethyl pyridine, 2-pyridin-3-yl ethanamine, 3-aminoethylpyridine, 3-pyridineethanamine, 2-pyridin-3-yl ethan-1-amine, 2-3-pyridyl ethylamine, 3-pyridineethaneamine, 3-pyridine ethanamine, 2-pyridin-3-yl-ethylamine, 2-3-pyridinyl ethanamine
PubChem CID 854051
SMILES [NH3+]CCC1=CC=CN=C1

Specifications

Boiling Point 119°C to 120°C (19 mmHg)
Odor Amine-like
Refractive Index 1.5518
Quantity 1 g
UN Number UN2735
Beilstein 111288
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
IUPAC Name 2-(pyridin-3-yl)ethan-1-aminium
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 3-(2-Aminoethyl)pyridine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only