missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)-5-bromoindole, 97%

Catalog No. AAH63653MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63653MD 250 mg
AAH6365303 1 g
AAH6365306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63653MD Supplier Thermo Scientific Chemicals Supplier No. H63653MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3610-42-2 | C10H11BrN2 | 239.116 g/mol

N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 3610-42-2
Molecular Formula C10H11BrN2
Molecular Weight (g/mol) 239.116
MDL Number MFCD00130169
InChI Key CGHUQJRRADEHTQ-UHFFFAOYSA-N
PubChem CID 77158
IUPAC Name 2-(5-bromo-1H-indol-3-yl)ethanamine
SMILES C1=CC2=C(C=C1Br)C(=CN2)CCN

Specifications

Quantity 250 mg
Synonym 5-bromotryptamine, 2-5-bromo-1h-indol-3-yl ethanamine, 5-bromo-1h-indole-3-ethylamine, 1h-indole-3-ethanamine,5-bromo, 2-5-bromo-1h-indol-3-yl ethan-1-amine, 1h-indole-3-ethanamine, 5-bromo, 2-5-bromo-1h-indol-3-yl-ethylamine, 5-bromo-tryptamine, 5-br-t, 3-2-aminoethyl-5-bromoindole
Formula Weight 239.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(2-Aminoethyl)-5-bromoindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-779-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only