CAS RN 3610-42-2
IUPAC Name:
2-(5-bromo-1H-indol-3-yl)ethan-1-amine
Synonyms:
2-(5-bromo-1H-indol-3-yl)ethanamine
Molecular Weight (g/mol):
239.12
Molecular Formula:
C10H11BrN2
InChi Key:
CGHUQJRRADEHTQ-UHFFFAOYSA-N
SMILES:
NCCC1=CNC2=CC=C(Br)C=C12
1
–
1
of
1
results
1
Promotions Available
3-(2-Aminoethyl)-5-bromoindole, 97%
3-(2-Aminoethyl)-5-bromoindole, 97%
CAS: 3610-42-2 Molecular Formula: C10H11BrN2 Molecular Weight (g/mol): 239.116 MDL Number: MFCD00130169 InChI Key: CGHUQJRRADEHTQ-UHFFFAOYSA-N PubChem CID: 77158 IUPAC Name: 2-(5-bromo-1H-indol-3-yl)ethanamine SMILES: C1=CC2=C(C=C1Br)C(=CN2)CCN