missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)-1-Boc-azetidine, 95%

CAS: 898271-20-0 | C10H20N2O2 | 200.282 g/mol

Supplier:  Thermo Scientific Chemicals H6046903

Quantity 1 g

 View more versions of this product

Catalog No. AAH6046903


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

898271-20-0
200.282
XQNQFOKZUQWWDO-UHFFFAOYSA-N
44630603
CC(C)(C)OC(=O)N1CC(C1)CCN
C10H20N2O2
MFCD09878606
1-boc-3-aminoethylazetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, 1-boc-3-aminoethyl azetidine, 1-n-boc-3-aminoethyl azetidine, 1-boc-3-2-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate
tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate

Specifications

3-(2-Aminoethyl)-1-Boc-azetidine
C10H20N2O2
1-boc-3-aminoethylazetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, 1-boc-3-aminoethyl azetidine, 1-n-boc-3-aminoethyl azetidine, 1-boc-3-2-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate
CC(C)(C)OC(=O)N1CC(C1)CCN
1 g
44630603
95%
898271-20-0
MFCD09878606
XQNQFOKZUQWWDO-UHFFFAOYSA-N
tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate
200.282
200.28
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions