accessibility menu, dialog, popup

UserName

CAS RN 898271-20-0

CH3H3CCH3OONNH2

CAS RN 898271-20-0

IUPAC Name: tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate
Synonyms: tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: XQNQFOKZUQWWDO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(CCN)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)