missing translation for 'onlineSavingsMsg'
Learn More

3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl Chloride 98.0+%, TCI America™

Catalog No. D47765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47765G 5 g
D477625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47765G Supplier TCI America Supplier No. D47765G

Chemical Identifiers

CAS 4462-55-9
Molecular Formula C11H6Cl3NO2
Molecular Weight (g/mol) 290.524
MDL Number MFCD00052556
InChI Key IZQGELJKDARDMZ-UHFFFAOYSA-N
Synonym 3-2,6-dichlorophenyl-5-methylisoxazole-4-carbonyl chloride, dcimc chloride, 3-2,6-dichlorophenyl-5-methyl-1,2-oxazole-4-carbonyl chloride, 3-2,6-dichlorophenyl-5-methylisoxazole-4-carbonylchloride, 4-isoxazolecarbonyl chloride, 3-2,6-dichlorophenyl-5-methyl, 3-2,6-dichlorophenyl-5-methylisoxazole-4-carboxylic acid chloride, 3-2,6-dichlorophenyl-5-methyl isoxazole-4-carbonyl chloride, pubchem8683, dsstox_cid_31536, dsstox_rid_97421
PubChem CID 78212
IUPAC Name 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)Cl

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 5 g
Formula Weight 290.52
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl Chloride
EINECSNumber (9)-0578
TSCA Yes