accessibility menu, dialog, popup

UserName

CH3ONClOClCl(?)

CAS RN 4462-55-9

IUPAC Name: 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl chloride
Synonyms: 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl chloride
Molecular Weight (g/mol): 290.52
Molecular Formula: C11H6Cl3NO2
InChi Key: IZQGELJKDARDMZ-UHFFFAOYSA-N
SMILES: CC1=C(C(Cl)=O)C(=NO1)C1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results