missing translation for 'onlineSavingsMsg'
Learn More

3-(2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate

Catalog No. p-7080141
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4653906 5 g
AA4653914 25 g
AA4653922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4653906 Supplier Thermo Scientific Chemicals Supplier No. 4653906
Only null left

CAS: 903876-45-9 | C19H30O2 | 290.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 903876-45-9
Molecular Formula C19H30O2
Molecular Weight (g/mol) 290.45
MDL Number MFCD22380703
InChI Key CDBRNGRSVNBVLJ-UHFFFAOYNA-N
Synonym 3-5,5,6-trimethylbicyclo 2.2.1 heptan-2-yl cyclohexyl prop-2-enoate

Specifications

Color Brown
Flash Point >149°C (300°F)
Quantity 5 g
Formula Weight 290.45
Physical Form Liquid
Chemical Name or Material 3- (2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only