missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-1-ylmethyl)benzylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC56113CB

Quantity 250mg

 View more versions of this product

Catalog No. CC56113CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

562803-76-3
187.246
XKFHMAZBNKKNMT-UHFFFAOYSA-N
16640564
C1=CC(=CC(=C1)CN)CN2C=CC=N2
C11H13N3
MFCD08572164
3-1h-pyrazol-1-ylmethyl benzylamine, 3-1h-pyrazol-1-ylmethyl phenyl methanamine, 1-3-aminomethyl benzyl-1h-pyrazole, 3-pyrazol-1-ylmethyl phenyl methanamine, 1-3-pyrazol-1-ylmethyl phenyl methanamine, 3-pyrazolylmethyl phenyl methylamine, 3-1h-pyrazol-1-yl methyl benzylamine, 3-1h-pyrazole-1-ylmethyl benzenemethanamine, benzenemethanamine,3-1h-pyrazol-1-ylmethyl, 3-1h-pyrazol-1-yl methyl phenyl methanamine
[3-(pyrazol-1-ylmethyl)phenyl]methanamine

Specifications

3-(1H-Pyrazol-1-ylmethyl)benzylamine
89.5
90%
C11H13N3
MFCD08572164
XKFHMAZBNKKNMT-UHFFFAOYSA-N
[3-(pyrazol-1-ylmethyl)phenyl]methanamine
16640564
90%
562803-76-3
100.0
Amber glass bottle
250mg
3-1h-pyrazol-1-ylmethyl benzylamine, 3-1h-pyrazol-1-ylmethyl phenyl methanamine, 1-3-aminomethyl benzyl-1h-pyrazole, 3-pyrazol-1-ylmethyl phenyl methanamine, 1-3-pyrazol-1-ylmethyl phenyl methanamine, 3-pyrazolylmethyl phenyl methylamine, 3-1h-pyrazol-1-yl methyl benzylamine, 3-1h-pyrazole-1-ylmethyl benzenemethanamine, benzenemethanamine,3-1h-pyrazol-1-ylmethyl, 3-1h-pyrazol-1-yl methyl phenyl methanamine
C1=CC(=CC(=C1)CN)CN2C=CC=N2
187.246
187.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions