accessibility menu, dialog, popup

UserName

H2NNN

CAS RN 562803-76-3

IUPAC Name: 1-{3-[(1H-pyrazol-1-yl)methyl]phenyl}methanamine
Synonyms: 1-[3-(pyrazol-1-ylmethyl)phenyl]methanamine
Molecular Weight (g/mol): 187.25
Molecular Formula: C11H13N3
InChi Key: XKFHMAZBNKKNMT-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(CN2C=CC=N2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)