missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1h-1,2,4-Triazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC45404DA
Specifications
| 3-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde | |
| 97% | |
| 1g | |
| IBVURYHFXBPNCW-UHFFFAOYSA-N | |
| 3-(1,2,4-triazol-1-ylmethyl)benzaldehyde | |
| 22360585 | |
| 97% |
| 876316-30-2 | |
| C10H9N3O | |
| 3-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazol-1-ylmethyl benzaldehyde, 3-1h-1,2,4-triazol-1-yl methyl benzaldehyde, 3-1,2,4-triazolylmethyl benzaldehyde | |
| C1=CC(=CC(=C1)CN2C=NC=N2)C=O | |
| 187.202 | |
| 187.2 |
Chemical Identifiers
| 876316-30-2 | |
| 187.202 | |
| 3-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazol-1-ylmethyl benzaldehyde, 3-1h-1,2,4-triazol-1-yl methyl benzaldehyde, 3-1,2,4-triazolylmethyl benzaldehyde | |
| 3-(1,2,4-triazol-1-ylmethyl)benzaldehyde |
| C10H9N3O | |
| IBVURYHFXBPNCW-UHFFFAOYSA-N | |
| 22360585 | |
| C1=CC(=CC(=C1)CN2C=NC=N2)C=O |