missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Piperazinylmethyl)indole, 95%
CAS: 114746-66-6 | C13H17N3 | 215.3 g/mol
$160.12 - $1546.41
Chemical Identifiers
| CAS | 114746-66-6 |
|---|---|
| Molecular Formula | C13H17N3 |
| Molecular Weight (g/mol) | 215.3 |
| InChI Key | ZTNAIHGOFCMOPJ-UHFFFAOYSA-N |
| Synonym | 3-piperazin-1-ylmethyl-1h-indole, 1h-indole,3-1-piperazinylmethyl, acmc-1c6nr, 1-1h-indole-3-ylmethyl piperazine, 1h-indole, 3-1-piperazinylmethyl-, hydrochloride |
| PubChem CID | 17880880 |
| IUPAC Name | 3-(piperazin-1-ylmethyl)-1H-indole |
| SMILES | C1CN(CCN1)CC2=CNC3=CC=CC=C32 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH63402MD
|
Thermo Scientific Chemicals
H63402MD |
250 mg |
Each for $160.12
|
|
|||||
|
AAH6340203
|
Thermo Scientific Chemicals
H6340203 |
1 g |
Each for $374.25
|
|
|||||
|
AAH6340206
|
Thermo Scientific Chemicals
H6340206 |
5 g |
Each for $1,546.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 114746-66-6 | |
| 215.3 | |
| 3-piperazin-1-ylmethyl-1h-indole, 1h-indole,3-1-piperazinylmethyl, acmc-1c6nr, 1-1h-indole-3-ylmethyl piperazine, 1h-indole, 3-1-piperazinylmethyl-, hydrochloride | |
| 3-(piperazin-1-ylmethyl)-1H-indole |
| C13H17N3 | |
| ZTNAIHGOFCMOPJ-UHFFFAOYSA-N | |
| 17880880 | |
| C1CN(CCN1)CC2=CNC3=CC=CC=C32 |
Specifications
| 114746-66-6 | |
| 250 mg | |
| ZTNAIHGOFCMOPJ-UHFFFAOYSA-N | |
| 3-(piperazin-1-ylmethyl)-1H-indole | |
| 17880880 | |
| 95% | |
| 3-(1-Piperazinylmethyl)indole |
| C13H17N3 | |
| 3-piperazin-1-ylmethyl-1h-indole, 1h-indole,3-1-piperazinylmethyl, acmc-1c6nr, 1-1h-indole-3-ylmethyl piperazine, 1h-indole, 3-1-piperazinylmethyl-, hydrochloride | |
| C1CN(CCN1)CC2=CNC3=CC=CC=C32 | |
| 215.3 | |
| 215.29 | |
| Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only