missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Piperazinylmethyl)indole, 95%

CAS: 114746-66-6 | C13H17N3 | 215.3 g/mol

Supplier:  Thermo Scientific Chemicals H6340206

 View more versions of this product

Catalog No. AAH6340206


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(1-Piperazinylmethyl)indole
C13H17N3
3-piperazin-1-ylmethyl-1h-indole, 1h-indole,3-1-piperazinylmethyl, acmc-1c6nr, 1-1h-indole-3-ylmethyl piperazine, 1h-indole, 3-1-piperazinylmethyl-, hydrochloride
C1CN(CCN1)CC2=CNC3=CC=CC=C32
215.3
215.29
Solid
114746-66-6
5 g
ZTNAIHGOFCMOPJ-UHFFFAOYSA-N
3-(piperazin-1-ylmethyl)-1H-indole
17880880
95%

Chemical Identifiers

114746-66-6
215.3
3-piperazin-1-ylmethyl-1h-indole, 1h-indole,3-1-piperazinylmethyl, acmc-1c6nr, 1-1h-indole-3-ylmethyl piperazine, 1h-indole, 3-1-piperazinylmethyl-, hydrochloride
3-(piperazin-1-ylmethyl)-1H-indole
C13H17N3
ZTNAIHGOFCMOPJ-UHFFFAOYSA-N
17880880
C1CN(CCN1)CC2=CNC3=CC=CC=C32
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only