missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Piperazinyl)phenol, 97%

Catalog No. AAL1870214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1870214 25 g
AAL1870206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1870214 Supplier Thermo Scientific Chemicals Supplier No. L1870214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59817-32-2 | C10H14N2O | 178.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59817-32-2
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD00052896
InChI Key AYGYICRITMSJOC-UHFFFAOYSA-N
Synonym 3-piperazin-1-yl phenol, 1-3-hydroxyphenyl piperazine, 3-1-piperazinyl phenol, n-3-hydroxyphenyl piperazine, 3-piperazin-1-yl-phenol, 1-3-hydroxyphenyl-piperazine, phenol, 3-1-piperazinyl, 3-piperazinylphenol, 3-hydroxyphenylpiperazine
PubChem CID 2736597
SMILES OC1=CC=CC(=C1)N1CCNCC1

Specifications

Melting Point 205°C to 208°C
Quantity 25 g
UN Number UN3263
Beilstein 4248583
Sensitivity Air sensitive
IUPAC Name 3-piperazin-1-ylphenol
Formula Weight 178.24
Percent Purity 98%
Chemical Name or Material 3-(1-Piperazinyl)phenol
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only