missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Piperazinyl)-1,2-benzisothiazole 98.0+%, TCI America™
$115.51 - $338.84
Chemical Identifiers
| CAS | 87691-87-0 |
|---|---|
| Molecular Formula | C11H13N3S |
| Molecular Weight (g/mol) | 219.306 |
| MDL Number | MFCD04117970 |
| InChI Key | KRDOFMHJLWKXIU-UHFFFAOYSA-N |
| PubChem CID | 2772144 |
| IUPAC Name | 3-piperazin-1-yl-1,2-benzothiazole |
| SMILES | C1CN(CCN1)C2=NSC3=CC=CC=C32 |
Chemical Identifiers
| 87691-87-0 | |
| 219.306 | |
| KRDOFMHJLWKXIU-UHFFFAOYSA-N | |
| 3-piperazin-1-yl-1,2-benzothiazole |
| C11H13N3S | |
| MFCD04117970 | |
| 2772144 | |
| C1CN(CCN1)C2=NSC3=CC=CC=C32 |
Specifications
| 87691-87-0 | |
| Yellow | |
| MFCD04117970 | |
| KRDOFMHJLWKXIU-UHFFFAOYSA-N | |
| 3-piperazin-1-yl-1,2-benzothiazole | |
| 2772144 | |
| ≥98.0% (GC,T) | |
| 3-(1-Piperazinyl)-1,2-benzisothiazole |
| 91°C | |
| C11H13N3S | |
| 1 g | |
| C1CN(CCN1)C2=NSC3=CC=CC=C32 | |
| 219.306 | |
| 219.31 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator