missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Piperazinyl)-1,2-benzisothiazole 98.0+%, TCI America™

Catalog No. P20641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20641G 1 g
P20645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20641G Supplier TCI America Supplier No. P20641G

Chemical Identifiers

CAS 87691-87-0
Molecular Formula C11H13N3S
Molecular Weight (g/mol) 219.306
MDL Number MFCD04117970
InChI Key KRDOFMHJLWKXIU-UHFFFAOYSA-N
PubChem CID 2772144
IUPAC Name 3-piperazin-1-yl-1,2-benzothiazole
SMILES C1CN(CCN1)C2=NSC3=CC=CC=C32

Specifications

Melting Point 91°C
Color Yellow
Quantity 1 g
Formula Weight 219.31
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(1-Piperazinyl)-1,2-benzisothiazole
TSCA No
Recommended Storage Refrigerator