missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Naphthyl)-D-alanine Hydrochloride 98.0+%, TCI America™
$61.61 - $300.33
Chemical Identifiers
| CAS | 122745-09-9 |
|---|---|
| Molecular Formula | C13H14ClNO2 |
| Molecular Weight (g/mol) | 251.71 |
| InChI Key | BKQQPCDQZZTLSE-UTONKHPSSA-N |
| Synonym | (R)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride |
| PubChem CID | 44629773 |
| IUPAC Name | (2R)-2-amino-3-naphthalen-1-ylpropanoic acid;hydrochloride |
| SMILES | C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)N.Cl |
Chemical Identifiers
| 122745-09-9 | |
| 251.71 | |
| (R)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride | |
| (2R)-2-amino-3-naphthalen-1-ylpropanoic acid;hydrochloride |
| C13H14ClNO2 | |
| BKQQPCDQZZTLSE-UTONKHPSSA-N | |
| 44629773 | |
| C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)N.Cl |
Specifications
| 122745-09-9 | |
| C13H14ClNO2 | |
| (R)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride | |
| C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)N.Cl | |
| 251.71 | |
| 251.71 | |
| Crystalline Powder |
| White | |
| 1 g | |
| BKQQPCDQZZTLSE-UTONKHPSSA-N | |
| (2R)-2-amino-3-naphthalen-1-ylpropanoic acid;hydrochloride | |
| 44629773 | |
| ≥98.0% (T) | |
| 3-(1-Naphthyl)-D-alanine Hydrochloride |
Safety and Handling
TSCA : No