accessibility menu, dialog, popup

UserName

CAS RN 122745-09-9

OHONH2HCl

CAS RN 122745-09-9

IUPAC Name: 2-amino-3-(naphthalen-1-yl)propanoic acid hydrochloride
Synonyms: alphanaphthylalanine hydrochloride
Molecular Weight (g/mol): 251.71
Molecular Formula: C13H14ClNO2
InChi Key: BKQQPCDQZZTLSE-UHFFFAOYNA-N
SMILES: Cl.NC(CC1=CC=CC2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results