missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)-D-alanine Hydrochloride 98.0+%, TCI America™

Catalog No. N07725G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07725G 5 g
N07721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07725G Supplier TCI America Supplier No. N07725G
Only null left

Chemical Identifiers

CAS 122745-09-9
Molecular Formula C13H14ClNO2
Molecular Weight (g/mol) 251.71
InChI Key BKQQPCDQZZTLSE-UTONKHPSSA-N
Synonym (R)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride
PubChem CID 44629773
IUPAC Name (2R)-2-amino-3-naphthalen-1-ylpropanoic acid;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)N.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 251.71
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(1-Naphthyl)-D-alanine Hydrochloride